siRNA Details
Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144867.2 | NP_659116.1 | 3, 4 | 371 | ||
GenBank mRNA | AK038905.1 | - | 3, 4 | 330 | ||
AK143220.1 | - | 3, 4 | 355 | |||
BC019561.1 | - | 3, 4 | 320 | 4459623 | ||
BY741062.1 | - | 3, 4 | 372 |

More Information
Other Species Targeted

