Search
Search

Fads1| Gene Symbol: Fads1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146094.2 | NP_666206.1 | 6 | 946 | ||
| GenBank mRNA | AB072976.1 | - | 6 | 861 | ||
| AF061504.1 | - | 6 | 848 | |||
| AK033308.1 | - | 4 | 303 | |||
| AK083959.1 | - | 6 | 808 | |||
| AK154367.1 | - | 6 | 896 | |||
| AK185930.1 | - | 2 | 95 | |||
| AK192444.1 | - | 2 | 95 | |||
| AK193280.1 | - | 2 | 95 | |||
| AK195832.1 | - | 168 | ||||
| AK198806.1 | - | 2 | 95 | |||
| AK213965.1 | - | 2 | 96 | |||
| BC022139.1 | - | 6 | 891 | 5100199 | ||
| BC026831.1 | - | 6 | 913 | 4239695 | ||
| BC026848.1 | - | 6 | 886 | 4196030 | ||
| BC063053.1 | - | 6 | 806 | 5696722 | ||
| KF987079.1 | - | 6 | 830 | |||