Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 12 | 1276 | ||
GenBank mRNA | AK034591.1 | - | 12 | 1171 | ||
AK079849.1 | - | 12 | 1213 | |||
AK083401.1 | - | 12 | 1190 | |||
AK083537.1 | - | 12 | 1197 | |||
AK087134.1 | - | 12 | 1252 | |||
AK088247.1 | - | 12 | 1193 | |||
AK134406.1 | - | 12 | 1167 | |||
AK165243.1 | - | 12 | 1191 | |||
AK166118.1 | - | 12 | 1191 | |||
AK169523.1 | - | 12 | 1228 | |||
BC018191.1 | - | 12 | 1177 | 3994248 | ||
BC025501.1 | - | 12 | 1223 | 5321013 | ||
BC026604.1 | - | 11 | 1730 |