siRNA Details
Gene Symbol: Ano6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253813.1 | NP_001240742.1 | 4, 5 | 499 | ||
NM_175344.4 | NP_780553.2 | 3, 4 | 436 | |||
GenBank mRNA | AK089300.1 | - | 3, 4 | 347 | ||
AK159411.1 | - | 4, 5 | 450 | |||
BC060732.1 | - | 3, 4 | 436 | 6409332 |

More Information
Other Species Targeted

