Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206924.2 | NP_996807.1 | 5 | 872 | ||
GenBank mRNA | AB016490.1 | - | 5 | 817 | ||
AK004469.1 | - | 5 | 784 | |||
AK089048.1 | - | 5 | 784 | |||
AK146337.1 | - | 5 | 873 | |||
BC008139.1 | - | 5 | 836 | 3582930 | ||
BG077727.2 | - | 4 | 438 | |||
BY617278.1 | - | 1 | 184 |