Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207223.1 | NP_997106.1 | 5, 6 | 867 | ||
GenBank mRNA | AK147505.1 | - | 5, 6 | 921 | ||
AK159055.1 | - | 3, 4 | 142 | |||
BC025556.1 | - | 5, 6 | 441 | |||
BC067016.1 | - | 5, 6 | 867 | 6827624 | ||
BC098196.1 | - | 5, 6 | 443 | 6847993 |