Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301781.1 | NP_001288710.1 | 4 | 593 | ||
NM_207232.2 | NP_997115.1 | 3 | 410 | |||
GenBank mRNA | AK030415.1 | - | 3 | 403 | ||
AK038922.1 | - | 3 | 323 | |||
AK053480.1 | - | 3 | 248 | |||
AK132781.1 | - | 4 | 502 | |||
BC066081.1 | - | 3 | 311 | 30355705 |