Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253752.1 | NP_001240681.1 | 5 | 977 | ||
NM_001253753.1 | NP_001240682.1 | 5 | 1047 | |||
NM_144842.3 | NP_659091.3 | 5 | 1041 | |||
GenBank mRNA | AK036513.1 | - | 4 | 903 | ||
AK155333.1 | - | 5 | 959 | |||
AK156501.1 | - | 5 | 1049 | |||
BC027380.1 | - | 3 | 390 | |||
BC058644.1 | - | 4 | 838 | |||
BE982095.1 | - | 2 | 301 |