Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145220.2 | NP_660255.1 | 21 | 2593 | ||
GenBank mRNA | AK160075.1 | - | 21 | 2572 | ||
AK160105.1 | - | 21 | 2569 | |||
AK168403.1 | - | 21 | 2556 | |||
AK168748.1 | - | 21 | 2549 | |||
AK171512.1 | - | 21 | 2549 | |||
AY113706.1 | - | 21 | 2595 | |||
BC048906.1 | - | 21 | 2531 | 6475576 |