Search
Search

Efhb| Gene Symbol: Efhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172497.3 | NP_766085.2 | 1 | 274 | ||
| GenBank mRNA | AK029550.1 | - | 1 | 275 | ||
| AK167235.1 | - | 1 | 259 | |||
| BC132103.1 | - | 1 | 265 | 40130380 | ||
| BC137880.1 | - | 1 | 265 | 8860899 | ||