Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177093.3 | NP_796067.2 | 2, 3 | 710 | ||
GenBank mRNA | AK049274.1 | - | 2, 3 | 711 | ||
AK090204.1 | - | 2, 3 | 719 | |||
AK147818.1 | - | 2, 3 | 686 | |||
BC141007.1 | - | 2, 3 | 670 | 9055619 |