Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mysm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177239.2 | NP_796213.2 | 7, 8 | 533 | ||
GenBank mRNA | AK043802.1 | - | 7, 8 | 533 | ||
AK081684.1 | - | 7, 8 | 508 | |||
AK164103.1 | - | 7, 8 | 479 | |||
AK173300.1 | - | 4, 5 | 275 | |||
BC150946.1 | - | 7, 8 | 529 | 9087857 | ||
BC151172.1 | - | 7, 8 | 529 | 9088061 |