Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177321.2 | NP_796295.1 | 4 | 1003 | ||
GenBank mRNA | AF390177.2 | - | 4 | 1003 |