Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRRM1 SIRNA Assays ›
          Gene Symbol:
          SRRM1
          Gene Name:
          serine and arginine repetitive matrix 1
          Gene Aliases:
          160-KD, POP101, SRM160
          Chromosome Location:
          Chr.1: 24643012 - 24673281 on Build GRCh38
          UniGene ID:
          Hs.18192
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 16870

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRRM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303448.1 NP_001290377.1 2, 3 343
          NM_001303449.1 NP_001290378.1 2, 3 272
          NM_005839.3 NP_005830.2 2, 3 343
          XM_005245717.2 XP_005245774.1 2, 3 253
          XM_005245718.2 XP_005245775.1 2, 3 253
          XM_005245720.2 XP_005245777.1 2, 3 253
          XM_005245721.2 XP_005245778.1 2, 3 253
          XM_005245722.2 XP_005245779.1 2, 3 253
          XM_011540478.2 XP_011538780.1 2, 3 267
          XM_011540479.1 XP_011538781.1 2, 3 434
          XM_011540480.1 XP_011538782.1 2, 3 434
          XM_011540481.1 XP_011538783.1 2, 3 434
          XM_011540482.1 XP_011538784.1 2, 3 434
          XM_011540483.1 XP_011538785.1 2, 3 434
          XM_017000011.1 XP_016855500.1 2, 3 267
          XM_017000012.1 XP_016855501.1 2, 3 267
          XM_017000013.1 XP_016855502.1 2, 3 267
          XM_017000014.1 XP_016855503.1 2, 3 267
          XM_017000015.1 XP_016855504.1 2, 3 267
          XM_017000016.1 XP_016855505.1 2, 3 289
          XM_017000017.1 XP_016855506.1 2, 3 267
          XM_017000018.1 XP_016855507.1 2, 3 253
          XM_017000019.1 XP_016855508.1 2, 3 253
          XM_017000020.1 XP_016855509.1 2, 3 253
          XM_017000021.1 XP_016855510.1 2, 3 253
          XM_017000022.1 XP_016855511.1 2, 3 253
          XM_017000023.1 XP_016855512.1 2, 3 434
          XM_017000024.1 XP_016855513.1 2, 3 434
          XM_017000025.1 XP_016855514.1 2, 3 253
          XM_017000026.1 XP_016855515.1 2, 3 253
          XM_017000027.1 XP_016855516.1 2, 3 434
          XM_017000028.1 XP_016855517.1 2, 3 434
          XM_017000029.1 XP_016855518.1 2, 3 434
          XM_017000030.1 XP_016855519.1 2, 3 434
          XM_017000031.1 XP_016855520.1 2, 3 434
          XM_017000032.1 XP_016855521.1 2, 3 434
          XM_017000033.1 XP_016855522.1 2, 3 434
          XM_017000034.1 XP_016855523.1 2, 3 267
          GenBank mRNA AF048977.1 - 2, 3 124
          AF419855.1 - 2, 3 346
          AK302498.1 - 2, 3 162
          AK307419.1 - 2, 3 272
          BC036187.1 - 2, 3 171 4827738
          BG720378.1 - 2, 3 346
          DC353932.1 - 2, 3 252
          KU178512.1 - 2, 3 119
          KU178513.1 - 2, 3 119

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107986, XM_006239185, XM_006239183, XM_008764243, XM_008764242, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001130477, NM_016799, BC094322, AK008284, AK169762, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          termination of RNA polymerase II transcription
          RNA export from nucleus
          mRNA export from nucleus
          RNA splicing
          mRNA 3'-end processing
          DNA binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0