Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Opn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181753.4 | NP_861418.2 | 5 | 983 | ||
GenBank mRNA | AK039525.1 | - | 1 | 174 | ||
AY288426.1 | - | 5 | 970 | |||
AY318865.1 | - | 5 | 984 | |||
BC125611.1 | - | 5 | 1065 | 40130126 | ||
BC132010.1 | - | 5 | 1065 | 40130287 |
Set Membership: |
G-protein coupled receptor |