Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa5b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144953.1 | NP_001138425.1 | 23 | 4227 | ||
GenBank mRNA | AK045595.1 | - | 2 | 2217 | ||
BC110635.2 | - | 1 | 2041 | |||
BC125016.1 | - | 1 | 2043 |