Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144817.2 | NP_659066.1 | 13 | 2280 | ||
GenBank mRNA | AF428262.1 | - | 13 | 2228 | ||
BC021840.1 | - | 13 | 2229 | 4502479 |