Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 6 | 878 | ||
NM_001164194.1 | NP_001157666.1 | 6 | 842 | |||
NM_005817.4 | NP_005808.3 | 6 | 878 | |||
GenBank mRNA | AF051314.1 | - | 2 | 190 | ||
AF051315.1 | - | 2 | 166 | |||
AF055574.1 | - | 6 | 754 | |||
AF057140.1 | - | 6 | 775 | |||
AK122671.1 | - | 6 | 729 | |||
AK223054.1 | - | 6 | 808 | |||
AK225045.1 | - | 6 | 744 | |||
AK225400.1 | - | 6 | 759 | |||
AK226088.1 | - | 6 | 756 | |||
AK291104.1 | - | 6 | 835 | |||
BC001590.2 | - | 6 | 751 | 2987965 | ||
BC005818.2 | - | 6 | 767 | 3833411 | ||
BC007566.1 | - | 6 | 767 | 3028104 | ||
BC019278.1 | - | 6 | 767 | 2905275 | ||
BQ057849.1 | - | 6 | 765 | |||
BQ063091.1 | - | 5 | 732 | |||
BT007235.1 | - | 5 | 701 |