Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271443.1 | NP_001258372.1 | 5 | 291 | ||
NM_001271444.1 | NP_001258373.1 | 5 | 531 | |||
NM_144870.5 | NP_659119.2 | 5 | 534 | |||
GenBank mRNA | AK135069.1 | - | 5 | 292 | ||
AK149674.1 | - | 5 | 492 | |||
AK153182.1 | - | 5 | 478 | |||
AK153900.1 | - | 5 | 268 | |||
AK164901.1 | - | 5 | 268 | |||
AK167907.1 | - | 5 | 269 | |||
BC021616.1 | - | 5 | 252 | 5132866 | ||
BC086766.1 | - | 5 | 245 | 6820786 |