Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSDE1 SIRNA Assays ›
          Gene Symbol:
          Csde1
          Gene Name:
          cold shock domain containing E1, RNA binding
          Gene Aliases:
          AA960392, BC016898, D3Jfr1, mKIAA0885, unr
          Chromosome Location:
          Chr.3: 103020431 - 103058189 on Build GRCm38
          UniGene ID:
          Mm.277713
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 169559

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Csde1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161854.1 NP_001155326.1 19 2978
          NM_144901.3 NP_659150.1 20 3071
          GenBank mRNA AK122393.1 - 19 2888
          AK144097.1 - 19 2918
          AK145351.1 - 19 3085
          AK145428.1 - 19 2902
          AK145932.1 - 19 2895
          AK146586.1 - 19 2919
          AK146706.1 - 20 3003
          AK147012.1 - 19 2917
          AK150882.1 - 19 2896
          AK151638.1 - 19 2914
          AK151723.1 - 19 2918
          AK151970.1 - 19 2918
          AK152531.1 - 19 2914
          AK152777.1 - 15 2125
          AK153196.1 - 19 2895
          AK153263.1 - 15 2125
          AK154068.1 - 19 2903
          AK155751.1 - 20 2996
          AK159258.1 - 19 2911
          AK159863.1 - 19 2911
          AK168556.1 - 18 2715
          BC016898.1 - 19 2915 4224466
          BC024826.1 - 16 2198
          BC029058.1 - 14 1857 4947555
          BC029688.1 - 19 2914 5367783
          BC031404.1 - 19 2886 4953772
          BC036720.1 - 16 2204
          BC062097.1 - 20 2986 6833505

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001242891, NM_001242893, NM_001130523, NM_001242892, NM_007158, and others.
          This siRNA also targets the following transcripts in Rat: NM_054006, XM_003751853, FQ222050, FQ230008, X52311, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          nuclear-transcribed mRNA catabolic process, no-go decay
          nucleic acid binding
          DNA binding
          RNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-s7hs7:80/100.66.74.89:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0