Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRRM1 SIRNA Assays ›
          Gene Symbol:
          SRRM1
          Gene Name:
          serine and arginine repetitive matrix 1
          Gene Aliases:
          160-KD, POP101, SRM160
          Chromosome Location:
          Chr.1: 24643012 - 24673281 on Build GRCh38
          UniGene ID:
          Hs.18192
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 16966

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRRM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303448.1 NP_001290377.1 3 370
          NM_001303449.1 NP_001290378.1 3 299
          NM_005839.3 NP_005830.2 3 370
          XM_005245717.2 XP_005245774.1 3 280
          XM_005245718.2 XP_005245775.1 3 280
          XM_005245720.2 XP_005245777.1 3 280
          XM_005245721.2 XP_005245778.1 3 280
          XM_005245722.2 XP_005245779.1 3 280
          XM_011540478.2 XP_011538780.1 3 294
          XM_011540479.1 XP_011538781.1 3 461
          XM_011540480.1 XP_011538782.1 3 461
          XM_011540481.1 XP_011538783.1 3 461
          XM_011540482.1 XP_011538784.1 3 461
          XM_011540483.1 XP_011538785.1 3 461
          XM_017000011.1 XP_016855500.1 3 294
          XM_017000012.1 XP_016855501.1 3 294
          XM_017000013.1 XP_016855502.1 3 294
          XM_017000014.1 XP_016855503.1 3 294
          XM_017000015.1 XP_016855504.1 3 294
          XM_017000016.1 XP_016855505.1 3 316
          XM_017000017.1 XP_016855506.1 3 294
          XM_017000018.1 XP_016855507.1 3 280
          XM_017000019.1 XP_016855508.1 3 280
          XM_017000020.1 XP_016855509.1 3 280
          XM_017000021.1 XP_016855510.1 3 280
          XM_017000022.1 XP_016855511.1 3 280
          XM_017000023.1 XP_016855512.1 3 461
          XM_017000024.1 XP_016855513.1 3 461
          XM_017000025.1 XP_016855514.1 3 280
          XM_017000026.1 XP_016855515.1 3 280
          XM_017000027.1 XP_016855516.1 3 461
          XM_017000028.1 XP_016855517.1 3 461
          XM_017000029.1 XP_016855518.1 3 461
          XM_017000030.1 XP_016855519.1 3 461
          XM_017000031.1 XP_016855520.1 3 461
          XM_017000032.1 XP_016855521.1 3 461
          XM_017000033.1 XP_016855522.1 3 461
          XM_017000034.1 XP_016855523.1 3 294
          GenBank mRNA AF048977.1 - 3 151
          AF419855.1 - 3 373
          AK302498.1 - 3 189
          AK307419.1 - 3 299
          BC036187.1 - 3 198 4827738
          BG720378.1 - 3 373
          DC353932.1 - 3 279
          KU178512.1 - 3 146
          KU178513.1 - 3 146

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107986, XM_006239185, XM_006239183, XM_008764243, XM_008764242, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001130477, NM_016799, BC094322, AK008284, AK169762, and others.

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          termination of RNA polymerase II transcription
          RNA export from nucleus
          mRNA export from nucleus
          RNA splicing
          mRNA 3'-end processing
          DNA binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0