Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shisa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145463.5 | NP_663438.1 | 2 | 1393 | ||
GenBank mRNA | AB280737.1 | - | 2 | 1147 | ||
AK034905.1 | - | 2 | 1368 | |||
AK087512.1 | - | 2 | 1363 | |||
BC024118.1 | - | 2 | 829 | 5068886 | ||
BC057640.1 | - | 2 | 957 | 4018213 | ||
DQ342342.1 | - | 2 | 607 |