Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC003331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077237.1 | NP_001070705.1 | 9 | 1105 | ||
NM_145511.2 | NP_663486.2 | 10 | 1201 | |||
GenBank mRNA | AK152022.1 | - | 7 | 621 | ||
AK152955.1 | - | 7 | 622 | |||
BC003331.1 | - | 8 | 790 | 3256634 | ||
CX243768.1 | - | 6 | 449 | |||
DQ077804.2 | - | 9 | 815 |