Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLCN SIRNA Assays ›
          Gene Symbol:
          Flcn
          Gene Name:
          folliculin
          Gene Aliases:
          AU014660, B430214A04Rik, Bhd, FLCL
          Chromosome Location:
          Chr.11: 59791408 - 59810039 on Build GRCm38
          UniGene ID:
          Mm.339640
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 170087

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Flcn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271356.1 NP_001258285.1 4 791
          NM_001271357.1 NP_001258286.1 4 534
          NM_146018.2 NP_666130.1 4 831
          GenBank mRNA AK039106.1 - 4 618
          AK080933.1 - 4 616
          AK136071.1 - 4 522
          AK148216.1 - 4 585
          AK154040.1 - 4 813
          AK161460.1 - 4 563
          AK172245.1 - 4 590
          BC025820.1 - 4 565 5099493
          CJ052419.1 - 4 569

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          cell-cell junction assembly
          negative regulation of gene expression
          negative regulation of mitochondrion organization
          negative regulation of angiogenesis
          hemopoiesis
          negative regulation of cell growth
          negative regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          TOR signaling
          regulation of TOR signaling
          negative regulation of TOR signaling
          positive regulation of TOR signaling
          regulation of cytokinesis
          negative regulation of Rho protein signal transduction
          regulation of histone acetylation
          positive regulation of cell adhesion
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of protein localization to nucleus
          negative regulation of cell proliferation involved in kidney development
          negative regulation of cellular respiration
          negative regulation of mitochondrial DNA metabolic process
          negative regulation of muscle tissue development
          negative regulation of post-translational protein modification
          negative regulation of energy homeostasis
          regulation of pro-B cell differentiation
          negative regulation of ATP biosynthetic process
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0