Search
Search

B4galt7| Gene Symbol: B4galt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146045.1 | NP_666157.1 | 2, 3 | 445 | ||
| GenBank mRNA | AK050684.1 | - | 2, 3 | 392 | ||
| AK141739.1 | - | 2, 3 | 409 | |||
| AK146855.1 | - | 2, 3 | 492 | |||
| AK171600.1 | - | 2, 3 | 461 | |||
| BC027195.1 | - | 2, 3 | 445 | 4224150 | ||
| BC055703.1 | - | 2, 3 | 660 | 6403624 | ||