Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
1703
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
29
3609
NM_001242560.1
NP_001229489.1
30
3970
NM_004834.4
NP_004825.3
29
3760
NM_145686.3
NP_663719.2
30
4084
NM_145687.3
NP_663720.1
30
3528
XM_005264048.2
XP_005264105.1
32
4113
XM_005264050.3
XP_005264107.1
31
3975
XM_005264052.3
XP_005264109.1
31
3972
XM_005264057.3
XP_005264114.1
31
3906
XM_005264058.3
XP_005264115.1
30
3897
XM_005264059.3
XP_005264116.1
31
3813
XM_005264060.3
XP_005264117.1
30
3801
XM_005264062.3
XP_005264119.1
30
3744
XM_005264063.3
XP_005264120.1
30
3741
XM_005264064.3
XP_005264121.1
29
3735
XM_005264065.3
XP_005264122.1
30
3651
XM_005264066.3
XP_005264123.1
30
3648
XM_005264068.3
XP_005264125.1
29
3642
XM_005264069.2
XP_005264126.1
29
3588
XM_005264071.2
XP_005264128.1
28
3516
XM_006712865.2
XP_006712928.1
31
3882
XM_006712868.2
XP_006712931.1
30
3627
XM_006712869.2
XP_006712932.1
29
3528
XM_017005349.1
XP_016860838.1
32
4017
XM_017005350.1
XP_016860839.1
31
3951
XM_017005351.1
XP_016860840.1
30
3894
XM_017005352.1
XP_016860841.1
31
3882
XM_017005353.1
XP_016860842.1
31
3879
XM_017005354.1
XP_016860843.1
31
3858
XM_017005355.1
XP_016860844.1
30
3849
XM_017005356.1
XP_016860845.1
30
3804
XM_017005357.1
XP_016860846.1
31
3789
XM_017005358.1
XP_016860847.1
31
3786
XM_017005359.1
XP_016860848.1
30
3783
XM_017005360.1
XP_016860849.1
30
3780
XM_017005361.1
XP_016860850.1
30
3759
XM_017005362.1
XP_016860851.1
30
3756
XM_017005363.1
XP_016860852.1
30
3720
XM_017005364.1
XP_016860853.1
29
3708
XM_017005365.1
XP_016860854.1
30
3690
XM_017005366.1
XP_016860855.1
30
3690
XM_017005367.1
XP_016860856.1
30
3663
XM_017005368.1
XP_016860857.1
30
3594
XM_017005369.1
XP_016860858.1
29
3549
XM_017005370.1
XP_016860859.1
29
3525
XM_017005371.1
XP_016860860.1
28
3519
XM_017005372.1
XP_016860861.1
29
3459
XM_017005373.1
XP_016860862.1
28
3450
XM_017005374.1
XP_016860863.1
29
3435
XM_017005375.1
XP_016860864.1
28
3426
GenBank mRNA
AB013385.1
-
13
1516
AB014587.1
-
28
3412
AF096300.1
-
29
3461
AK001272.1
-
13
1671
AK025610.1
-
17
2285
AK074592.1
-
9
1277
AK295610.1
-
29
3754
AK296396.1
-
30
3666
AL137755.1
-
22
3066
AY212247.1
-
29
3604
AY834276.1
-
25
3013
EU794634.1
-
30
3578
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0