Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304990.1 | NP_001291919.1 | 3 | 2106 | ||
NM_005840.2 | NP_005831.1 | 2 | 2250 | |||
GenBank mRNA | AK094610.1 | - | 1 | 197 | ||
AK125483.1 | - | 3 | 1877 | |||
AK311508.1 | - | 2 | 1073 |