Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172148.1 | NP_742160.1 | 3 | 645 | ||
GenBank mRNA | AK141500.1 | - | 4 | 620 | ||
AK146227.1 | - | 4 | 593 | |||
BC028440.1 | - | 3 | 645 | 1265537 |