Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097623.1 | NP_001091092.1 | 6 | 1954 | ||
NM_172622.2 | NP_766210.1 | 3 | 1791 | |||
GenBank mRNA | AK035176.1 | - | 1 | 1561 | ||
AK042320.1 | - | 4 | 2095 | |||
AK046822.1 | - | 1 | 1432 | |||
AK154542.1 | - | 5 | 1967 | |||
AK155049.1 | - | 5 | 2068 | |||
AK155404.1 | - | 5 | 1965 | |||
AY081774.1 | - | 2 | 1704 | |||
BC059215.1 | - | 6 | 1954 | 6412734 | ||
BC108416.1 | - | 1 | 1414 | 5697521 |