Search
Search

Slc25a24| Gene Symbol: Slc25a24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172685.3 | NP_766273.1 | 10 | 3309 | ||
| GenBank mRNA | AK011425.1 | - | 1 | 919 | ||
| AK032806.1 | - | 10 | 3310 | |||
| AK146034.1 | - | 10 | 3308 | |||
| AK167778.1 | - | 10 | 3300 | |||
| AK168090.1 | - | 10 | 3300 | |||
| AK170598.1 | - | 10 | 3302 | |||
| BC055369.1 | - | 10 | 3298 | 30055307 | ||