Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199283.1 | NP_001186212.1 | 14 | 4031 | ||
NM_001199284.1 | NP_001186213.1 | 14 | 3813 | |||
NM_173388.2 | NP_775564.1 | 14 | 3776 | |||
GenBank mRNA | AK143773.1 | - | 14 | 3874 | ||
BC042513.1 | - | 14 | 3772 | 3964020 |