Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161851.2 | NP_001155323.1 | 6 | 1227 | ||
NM_001161852.2 | NP_001155324.1 | 5 | 1379 | |||
NM_173401.5 | NP_775577.2 | 6 | 1504 | |||
GenBank mRNA | AK077452.1 | - | 6 | 1290 | ||
AK170958.1 | - | 7 | 1271 | |||
AK177141.1 | - | 1 | 373 | |||
AK182933.1 | - | 1 | 294 | |||
AK184124.1 | - | 1 | 265 | |||
AK188052.1 | - | 1 | 270 | |||
AK188531.1 | - | 1 | 275 | |||
AK218373.1 | - | 1 | 38 | |||
AK218416.1 | - | 1 | 243 | |||
CA322196.1 | - | 3 | 683 |