Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290625.1 | NP_001277554.1 | 4 | 569 | ||
NM_001290626.1 | NP_001277555.1 | 3 | 567 | |||
NM_177345.4 | NP_796319.1 | 4 | 895 | |||
GenBank mRNA | AK042154.1 | - | 3 | 557 | ||
AK052045.1 | - | 4 | 563 | |||
AK132263.1 | - | 4 | 559 | |||
AK148428.1 | - | 4 | 884 | |||
AK178100.1 | - | 2 | 143 | |||
BC043296.1 | - | 4 | 794 | |||
BC090644.1 | - | 3 | 469 | 30629263 | ||
BC096618.1 | - | 4 | 868 | 6392317 |