Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290503.1 | NP_001277432.1 | 6 | 1423 | ||
NM_178694.4 | NP_848809.2 | 6 | 1462 | |||
GenBank mRNA | AK029290.1 | - | 6 | 1397 | ||
AK032153.1 | - | 6 | 1393 | |||
AK032318.1 | - | 5 | 1069 | |||
AK035712.1 | - | 2 | 2244 | |||
AK048853.1 | - | 6 | 1318 | |||
AK081355.1 | - | 6 | 1300 | |||
AK087046.1 | - | 6 | 1296 | |||
AK155401.1 | - | 6 | 1370 | |||
BC048924.1 | - | 6 | 1305 | 6312691 | ||
BC057021.1 | - | 6 | 1367 | 6825937 |