Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198108.2 | NP_932776.1 | 5 | 733 | ||
GenBank mRNA | AK154147.1 | - | 5 | 733 | ||
AK159230.1 | - | 5 | 732 | |||
AK187606.1 | - | 1 | 43 | |||
BC023055.1 | - | 5 | 711 | |||
BC057060.1 | - | 4 | 1109 | |||
CV563048.1 | - | 5 | 659 |