Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sertad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177794.1 | NP_001171265.1 | 4 | 2458 | ||
NM_198247.2 | NP_937890.2 | 4 | 2713 | |||
GenBank mRNA | AK029355.1 | - | 1 | 1244 | ||
AK047877.1 | - | 1 | 1458 | |||
BC060251.1 | - | 4 | 2682 | 5708574 |