Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007794.1 | NP_001007795.1 | 5 | 907 | ||
NM_006090.3 | NP_006081.1 | 5 | 869 | |||
XM_006710285.3 | XP_006710348.1 | 5 | 861 | |||
GenBank mRNA | AF068302.1 | - | 5 | 827 | ||
AF138862.1 | - | 4 | 737 | |||
AK023381.1 | - | 5 | 856 | |||
BC032610.2 | - | 6 | 869 | 5554219 | ||
BC049196.1 | - | 5 | 907 | 5226538 |