Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hivep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007772.2 | NP_031798.2 | 4 | 447 | ||
GenBank mRNA | AK039808.1 | - | 4 | 447 |