Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD200 SIRNA Assays ›
Gene Symbol:
CD200
Gene Name:
CD200 molecule
Gene Aliases:
MOX1, MOX2, MRC, OX-2
Chromosome Location:
Chr.3: 112332224 - 112362812 on Build GRCh38
UniGene ID:
Hs.79015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
17204
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD200
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004196.3
NP_001004196.2
4
520
NM_001318826.1
NP_001305755.1
3
445
NM_001318828.1
NP_001305757.1
2
363
NM_001318830.1
NP_001305759.1
2
363
NM_005944.6
NP_005935.4
3
445
XM_005247481.4
XP_005247538.1
4
694
XM_005247482.2
XP_005247539.1
3
324
XM_005247485.2
XP_005247542.1
3
635
XM_005247486.2
XP_005247543.1
2
132
XM_005247488.2
XP_005247545.1
2
242
XM_017006452.1
XP_016861941.1
4
1021
XM_017006453.1
XP_016861942.1
2
287
XM_017006454.1
XP_016861943.1
4
1010
XM_017006455.1
XP_016861944.1
3
645
XM_017006456.1
XP_016861945.1
4
371
XM_017006457.1
XP_016861946.1
3
324
XM_017006458.1
XP_016861947.1
4
1016
XM_017006459.1
XP_016861948.1
3
635
GenBank mRNA
AF063591.1
-
3
236
AK074972.1
-
3
210
AK129797.1
-
2
132
AK293399.1
-
2
132
AK297194.1
-
4
311
AK312563.1
-
3
326
AL832529.1
-
3
230
AY603771.1
-
2
179
BC022522.1
-
3
238
4794050
BC031103.1
-
3
214
5301702
BI547787.1
-
4
316
DA544134.1
-
3
323
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
negative regulation of macrophage activation
regulation of immune response
receptor activity
receptor binding
protein binding
protein homodimerization activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0