Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005981.3 | NP_005972.1 | 6 | 1019 | ||
XM_005269074.3 | XP_005269131.2 | 6 | 1944 | |||
XM_017019793.1 | XP_016875282.1 | 4 | 748 | |||
GenBank mRNA | AK090502.1 | - | 4 | 1422 | ||
AK294128.1 | - | 6 | 972 | |||
BC010377.1 | - | 6 | 972 | 4294627 | ||
BX647402.1 | - | 4 | 745 | |||
U01160.1 | - | 6 | 967 |