Search
Search

Tars| Gene Symbol: Tars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033074.3 | NP_149065.2 | 2, 3 | 251 | ||
| GenBank mRNA | AK010256.1 | - | 2, 3 | 218 | ||
| AK011146.2 | - | 2, 3 | 252 | |||
| AK031064.1 | - | 2, 3 | 215 | |||
| AK152945.1 | - | 2, 3 | 212 | |||
| AK153314.1 | - | 2, 3 | 223 | |||
| AK166693.1 | - | 2, 3 | 218 | |||
| AK167597.1 | - | 2, 3 | 260 | |||
| AK168969.1 | - | 2, 3 | 211 | |||
| AK197272.1 | - | 1, 2 | 57 | |||
| AK206659.1 | - | 1, 2 | 57 | |||
| AK210237.1 | - | 1, 2 | 57 | |||
| BC055371.1 | - | 2, 3 | 237 | 30060749 | ||
| BC060176.1 | - | 194 | ||||