Search
Search

B3gnt5| Gene Symbol: B3gnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159407.1 | NP_001152879.1 | 6 | 1275 | ||
| NM_001159408.1 | NP_001152880.1 | 5 | 1072 | |||
| NM_054052.3 | NP_473393.2 | 4 | 951 | |||
| GenBank mRNA | AB059620.1 | - | 1 | 124 | ||
| AK032911.1 | - | 4 | 865 | |||
| AK037166.1 | - | 5 | 1099 | |||
| AK048010.1 | - | 4 | 836 | |||
| AK083839.1 | - | 4 | 951 | |||
| AY029203.1 | - | 3 | 766 | |||
| BC063076.1 | - | 4 | 787 | 6848071 | ||