Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133225.3 | NP_573488.2 | 8 | 2156 | ||
GenBank mRNA | AK018392.1 | - | 1 | 337 | ||
AK030371.1 | - | 8 | 2157 | |||
AK168799.1 | - | 8 | 2130 | |||
BC060602.1 | - | 8 | 2111 | 5706182 |