Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133251.1 | NP_573514.1 | 4 | 519 | ||
GenBank mRNA | AB036708.1 | - | 4 | 519 | ||
AK146118.1 | - | 3 | 390 | |||
BC144928.1 | - | 4 | 467 | 9053476 | ||
BC145949.1 | - | 3 | 451 | 40131252 |