Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291039.1 | NP_001277968.1 | 13 | 1381 | ||
NM_138651.7 | NP_619592.1 | 13 | 1704 | |||
GenBank mRNA | AK036328.1 | - | 13 | 1555 | ||
AK049257.1 | - | 13 | 1382 | |||
AK147541.1 | - | 13 | 1596 | |||
AK166001.1 | - | 13 | 1555 | |||
AY159802.1 | - | 13 | 1627 | |||
BC003852.1 | - | 13 | 1610 | 3600825 | ||
BC069879.1 | - | 13 | 1583 | 30439069 |