Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291039.1 | NP_001277968.1 | 7 | 604 | ||
NM_138651.7 | NP_619592.1 | 7 | 927 | |||
GenBank mRNA | AK036328.1 | - | 7 | 778 | ||
AK049257.1 | - | 7 | 605 | |||
AK147541.1 | - | 7 | 819 | |||
AK166001.1 | - | 7 | 778 | |||
AY159802.1 | - | 7 | 850 | |||
BC003852.1 | - | 7 | 833 | 3600825 | ||
BC069879.1 | - | 7 | 806 | 30439069 |