Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138748.5 | NP_620087.3 | 6 | 791 | ||
GenBank mRNA | AK031591.1 | - | 6 | 792 | ||
AK046288.1 | - | 6 | 595 | |||
AK049184.1 | - | 6 | 782 | |||
AK150029.1 | - | 6 | 786 | |||
AK160980.1 | - | 6 | 646 | |||
AY035997.1 | - | 6 | 644 | |||
BC006626.1 | - | 6 | 707 | 3158086 | ||
BC052852.1 | - | 6 | 717 | 30058414 |