Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmem63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144794.2 | NP_659043.1 | 11 | 906 | ||
GenBank mRNA | AK049360.1 | - | 11 | 900 | ||
AK144899.1 | - | 11 | 959 | |||
AK170559.1 | - | 11 | 922 | |||
AK173027.1 | - | 11 | 899 | |||
BC014795.1 | - | 11 | 879 | 3961992 | ||
BC019442.1 | - | 11 | 906 | 4036849 |