Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PROM1 SIRNA Assays ›
          Gene Symbol:
          PROM1
          Gene Name:
          prominin 1
          Gene Aliases:
          AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
          Chromosome Location:
          Chr.4: 15968226 - 16084100 on Build GRCh38
          UniGene ID:
          Hs.614734
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 17327

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PROM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145847.1 NP_001139319.1 17 2430
          NM_001145848.1 NP_001139320.1 17 2146
          NM_001145849.1 NP_001139321.1 17 2150
          NM_001145850.1 NP_001139322.1 17 2150
          NM_001145851.1 NP_001139323.1 16 2123
          NM_001145852.1 NP_001139324.1 16 2123
          NM_006017.2 NP_006008.1 17 2150
          XM_005248195.4 XP_005248252.1 17 2246
          XM_005248196.4 XP_005248253.1 17 2246
          XM_006713974.3 XP_006714037.1 16 1751
          XM_011513890.1 XP_011512192.1 18 2232
          XM_011513892.2 XP_011512194.1 18 2383
          XM_011513893.2 XP_011512195.1 18 2457
          XM_011513894.2 XP_011512196.1 18 2542
          XM_011513895.2 XP_011512197.1 19 2658
          XM_011513896.2 XP_011512198.1 18 2287
          XM_011513897.2 XP_011512199.1 18 2474
          XM_011513898.2 XP_011512200.2 18 2232
          XM_011513899.2 XP_011512201.2 17 2246
          XM_011513900.2 XP_011512202.1 18 2232
          XM_011513902.2 XP_011512204.1 18 2232
          XM_011513903.2 XP_011512205.1 17 1781
          XM_017008799.1 XP_016864288.1 17 2276
          XM_017008800.1 XP_016864289.1 18 2232
          XM_017008801.1 XP_016864290.1 18 2232
          XM_017008802.1 XP_016864291.1 18 2324
          XM_017008803.1 XP_016864292.1 17 2221
          XM_017008804.1 XP_016864293.1 17 2291
          XM_017008805.1 XP_016864294.1 17 2164
          GenBank mRNA AF027208.1 - 17 1975
          AF117225.1 - 17 2191
          AF507034.1 - 16 1948
          AK027420.1 - 17 2169
          AK027422.1 - 17 2191
          AK075411.1 - 17 2146
          AK226033.1 - 17 2169
          AK314529.1 - 17 2190
          AY449689.1 - 16 1911
          AY449690.1 - 16 1911
          AY449691.1 - 16 1911
          AY449692.1 - 17 1938
          AY449693.1 - 17 1938
          BC012089.1 - 17 2146 4644690
          HQ628626.1 - 16 1911
          HQ628627.1 - 18 2167

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          retina layer formation
          photoreceptor cell maintenance
          retina morphogenesis in camera-type eye
          camera-type eye photoreceptor cell differentiation
          glomerular visceral epithelial cell differentiation
          glomerular parietal epithelial cell differentiation
          positive regulation of nephron tubule epithelial cell differentiation
          protein binding
          actinin binding
          cadherin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0