Search
Search

APBB3| Gene Symbol: APBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006051.3 | NP_006042.3 | 13 | 1919 | ||
| NM_133172.2 | NP_573418.2 | 12 | 1913 | |||
| NM_133173.2 | NP_573419.2 | 13 | 1898 | |||
| NM_133174.2 | NP_573420.2 | 12 | 1892 | |||
| GenBank mRNA | AB018247.1 | - | 13 | 1590 | ||
| AB049618.1 | - | 13 | 1782 | |||
| AB209839.1 | - | 13 | 1626 | |||
| AF224708.1 | - | 12 | 1574 | |||
| AF224709.1 | - | 13 | 1559 | |||
| AF224710.1 | - | 12 | 1553 | |||
| AF224711.1 | - | 13 | 1580 | |||
| AK074885.1 | - | 8 | 1825 | |||
| AK075280.1 | - | 13 | 1861 | |||
| AK125244.1 | - | 10 | 2551 | |||
| AK304824.1 | - | 9 | 1919 | |||
| BC013158.1 | - | 13 | 1898 | 3870148 | ||
| BC125187.1 | - | 13 | 1890 | 40122344 | ||